Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCTGTGAATGGGGGGTTTGGGA[C/T]AGCCGGGCTGGAGAAGGGATGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603186 MIM: 601962 MIM: 611439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAXX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAXX - death domain associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141969.1 | 1172 | Silent Mutation | CTA,CTG | L,L 706 | NP_001135441.1 | |
NM_001141970.1 | 1172 | Silent Mutation | CTA,CTG | L,L 718 | NP_001135442.1 | |
NM_001254717.1 | 1172 | Silent Mutation | CTA,CTG | L,L 631 | NP_001241646.1 | |
NM_001350.4 | 1172 | Silent Mutation | CTA,CTG | L,L 706 | NP_001341.1 | |
XM_005248860.3 | 1172 | Silent Mutation | CTA,CTG | L,L 338 | XP_005248917.1 |
TAPBP - TAP binding protein (tapasin) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB22 - zinc finger and BTB domain containing 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |