Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACATCCCATTTATGAGGTAGTC[C/T]GATTCCTCAGATGTAATGGGACGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176851 | ||||||||||||||||||||
Literature Links: |
LRP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRP11 - LDL receptor related protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032832.5 | 1797 | Silent Mutation | NP_116221.3 | |||
XM_011536194.2 | 1797 | Silent Mutation | XP_011534496.1 | |||
XM_017011380.1 | 1797 | Intron | XP_016866869.1 |
PCMT1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |