Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAACAGGAGAAGAAAATTAGACA[C/T]GATATCCTGACTGGGAACTGGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611831 | ||||||||||||||||||||
Literature Links: |
MRPL18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL18 - mitochondrial ribosomal protein L18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNLDC1 - PARN like, ribonuclease domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271862.1 | 514 | Silent Mutation | CAC,CAT | H,H 143 | NP_001258791.1 | |
NM_173516.2 | 514 | Silent Mutation | CAC,CAT | H,H 132 | NP_775787.1 | |
XM_011535491.2 | 514 | Silent Mutation | CAC,CAT | H,H 132 | XP_011533793.1 | |
XM_011535493.2 | 514 | Intron | XP_011533795.1 | |||
XM_017010312.1 | 514 | Silent Mutation | CAC,CAT | H,H 175 | XP_016865801.1 | |
XM_017010313.1 | 514 | Silent Mutation | CAC,CAT | H,H 137 | XP_016865802.1 | |
XM_017010314.1 | 514 | Silent Mutation | CAC,CAT | H,H 175 | XP_016865803.1 | |
XM_017010315.1 | 514 | Silent Mutation | CAC,CAT | H,H 105 | XP_016865804.1 | |
XM_017010316.1 | 514 | Silent Mutation | CAC,CAT | H,H 94 | XP_016865805.1 |