Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGCCGTACAGAGTGCGTCCTTG[A/C]CGCTTGAGCGCGTAAACCACATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142709 MIM: 602795 MIM: 602810 MIM: 602819 MIM: 602822 MIM: 602829 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H1A - histone cluster 1, H1a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2AB - histone cluster 1, H2ab | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3A - histone cluster 1, H3a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3B - histone cluster 1, H3b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4A - histone cluster 1, H4a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4B - histone cluster 1, H4b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003544.2 | Intron | NP_003535.1 |