Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610060 MIM: 612658 MIM: 607845 MIM: 609775 | ||||||||||||||||||||
Literature Links: |
LRRC73 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC73 - leucine rich repeat containing 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR1C - RNA polymerase I subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TJAP1 - tight junction associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPO5 - exportin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YIPF3 - Yip1 domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015388.3 | 1060 | Silent Mutation | CTG,TTG | L,L 281 | NP_056203.2 | |
XM_005248989.4 | 1060 | Silent Mutation | CTG,TTG | L,L 246 | XP_005249046.1 | |
XM_005248990.2 | 1060 | Silent Mutation | CTG,TTG | L,L 246 | XP_005249047.1 | |
XM_011514458.2 | 1060 | Silent Mutation | CTG,TTG | L,L 246 | XP_011512760.1 |