Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTGTTTTTTGCTTTTAGCATA[C/G]TAAACTCTCCTGTAATGAAAATGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613542 MIM: 607191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRF1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTRF1L - mitochondrial translational release factor 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGS17 - regulator of G-protein signaling 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012419.4 | 1445 | UTR 3 | NP_036551.3 | |||
XM_017010733.1 | 1445 | Intron | XP_016866222.1 |