Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGGGAATTGTAGCCAGCTGCC[A/G]TGAAATTATCTTTATATCTTTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHA7 - EPH receptor A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288629.1 | 3075 | Missense Mutation | ACG,ATG | T,M 940 | NP_001275558.1 | |
NM_001288630.1 | 3075 | Intron | NP_001275559.1 | |||
NM_004440.3 | 3075 | Missense Mutation | ACG,ATG | T,M 945 | NP_004431.1 | |
XM_005248669.2 | 3075 | Missense Mutation | ACG,ATG | T,M 941 | XP_005248726.1 | |
XM_005248671.2 | 3075 | Intron | XP_005248728.1 | |||
XM_017010365.1 | 3075 | Missense Mutation | ACG,ATG | T,M 936 | XP_016865854.1 | |
XM_017010366.1 | 3075 | Intron | XP_016865855.1 |