Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGCGGGACTCAGGTGAGCTCCG[A/G]AGGCTCCTCAACTCTTTGGAGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC12 - armadillo repeat containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286574.1 | 290 | Silent Mutation | CGA,CGG | R,R 71 | NP_001273503.1 | |
NM_001286576.1 | 290 | Silent Mutation | CGA,CGG | R,R 71 | NP_001273505.1 | |
NM_145028.4 | 290 | Silent Mutation | CGA,CGG | R,R 98 | NP_659465.2 | |
XM_011514381.2 | 290 | Silent Mutation | CGA,CGG | R,R 33 | XP_011512683.1 | |
XM_011514382.2 | 290 | UTR 5 | XP_011512684.1 | |||
XM_017010435.1 | 290 | Silent Mutation | CGA,CGG | R,R 98 | XP_016865924.1 |
FKBP5 - FK506 binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC285847 - uncharacterized LOC285847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |