Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAAGCAATGAAATGCCGAGGGC[A/G]GAGCCAAGAGCGACACTGGGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615353 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC28A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC28A - coiled-coil domain containing 28A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015439.2 | 167 | Silent Mutation | GCA,GCG | A,A 4 | NP_056254.1 | |
XM_011535728.2 | 167 | Silent Mutation | GCA,GCG | A,A 4 | XP_011534030.1 |
GVQW2 - GVQW motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242740.1 | 167 | Intron | NP_001229669.1 |