Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGGGAGTGGGATGGGGCTGAA[C/G]AGTGGTGATTTGGTGGCCCCGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609316 | ||||||||||||||||||||
Literature Links: |
TRIM31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM31 - tripartite motif containing 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007028.4 | 1171 | Missense Mutation | TCT,TGT | S,C 354 | NP_008959.3 | |
XM_011514264.1 | 1171 | Missense Mutation | TCT,TGT | S,C 353 | XP_011512566.1 | |
XM_011514265.1 | 1171 | Missense Mutation | TCT,TGT | S,C 354 | XP_011512567.1 | |
XM_011514266.1 | 1171 | Intron | XP_011512568.1 |
TRIM31-AS1 - TRIM31 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |