Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCCAAGGTAGCAGTCACTCAT[A/G]AAATATAATGTGTAGATATACCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614217 | ||||||||||||||||||||
Literature Links: |
ASCC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASCC3 - activating signal cointegrator 1 complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284271.1 | 7092 | Intron | NP_001271200.1 | |||
NM_006828.3 | 7092 | Silent Mutation | TTC,TTT | F,F 2161 | NP_006819.2 | |
NM_022091.4 | 7092 | Intron | NP_071374.1 | |||
XM_011535394.2 | 7092 | Silent Mutation | TTC,TTT | F,F 2166 | XP_011533696.1 | |
XM_011535395.2 | 7092 | Silent Mutation | TTC,TTT | F,F 2063 | XP_011533697.1 | |
XM_011535396.2 | 7092 | Silent Mutation | TTC,TTT | F,F 2063 | XP_011533698.1 | |
XM_017010205.1 | 7092 | Silent Mutation | TTC,TTT | F,F 2063 | XP_016865694.1 | |
XM_017010206.1 | 7092 | Silent Mutation | TTC,TTT | F,F 1688 | XP_016865695.1 |