Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGGAAGCCTACGTGATGGCCA[G/T]CGTGGACAACCCCCACGTGTGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 131550 | ||||||||||||||||||||
Literature Links: |
EGFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EGFR - epidermal growth factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005228.3 | 2549 | Missense Mutation | AGC,ATC | S,I 768 | NP_005219.2 | |
NM_201282.1 | 2549 | Intron | NP_958439.1 | |||
NM_201283.1 | 2549 | Intron | NP_958440.1 | |||
NM_201284.1 | 2549 | Intron | NP_958441.1 |
EGFR-AS1 - EGFR antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |