Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCCGGCCTCCCCTCCCCTCTC[C/T]CTGCTTGTGCTGCACAGGCTGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600859 MIM: 613635 MIM: 600259 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIMP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIMP2 - aminoacyl tRNA synthetase complex interacting multifunctional protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326606.1 | 395 | Silent Mutation | TCC,TCT | S,S 94 | NP_001313535.1 | |
NM_001326607.1 | 395 | Silent Mutation | TCC,TCT | S,S 65 | NP_001313536.1 | |
NM_001326609.1 | 395 | Silent Mutation | TCC,TCT | S,S 56 | NP_001313538.1 | |
NM_001326611.1 | 395 | Silent Mutation | TCC,TCT | S,S 56 | NP_001313540.1 | |
NM_006303.3 | 395 | Silent Mutation | TCC,TCT | S,S 134 | NP_006294.2 | |
XM_005249847.3 | 395 | Silent Mutation | TCC,TCT | S,S 127 | XP_005249904.1 | |
XM_005249848.2 | 395 | Silent Mutation | TCC,TCT | S,S 94 | XP_005249905.1 | |
XM_005249850.3 | 395 | Silent Mutation | TCC,TCT | S,S 56 | XP_005249907.1 | |
XM_006715772.2 | 395 | Silent Mutation | TCC,TCT | S,S 90 | XP_006715835.1 | |
XM_011515519.1 | 395 | Silent Mutation | TCC,TCT | S,S 105 | XP_011513821.1 |
EIF2AK1 - eukaryotic translation initiation factor 2 alpha kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMS2 - PMS1 homolog 2, mismatch repair system component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |