Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGCCTGTTGCAGCCATGGTGC[A/C]TTGCAGTTGCGTGTTGTTCAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610920 | ||||||||||||||||||||
Literature Links: |
GTPBP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTPBP10 - GTP binding protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042717.2 | 85 | Missense Mutation | CAT,CCT | H,P 3 | NP_001036182.1 | |
NM_033107.3 | 85 | Missense Mutation | CAT,CCT | H,P 3 | NP_149098.2 |
LOC101927446 - uncharacterized LOC101927446 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |