Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCGGAGGGTGTGCACACTTGC[C/T]GTGGACACCATCCCGTTGAAGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLXIPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLXIPL - MLX interacting protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032951.2 | 2412 | Silent Mutation | ACA,ACG | T,T 788 | NP_116569.1 | |
NM_032952.2 | 2412 | Silent Mutation | ACA,ACG | T,T 769 | NP_116570.1 | |
NM_032953.2 | 2412 | Silent Mutation | ACA,ACG | T,T 786 | NP_116571.1 | |
NM_032954.2 | 2412 | Silent Mutation | ACA,ACG | T,T 767 | NP_116572.1 | |
XM_011516277.1 | 2412 | Silent Mutation | ACA,ACG | T,T 853 | XP_011514579.1 | |
XM_011516278.1 | 2412 | Silent Mutation | ACA,ACG | T,T 852 | XP_011514580.1 | |
XM_011516279.1 | 2412 | Silent Mutation | ACA,ACG | T,T 835 | XP_011514581.1 | |
XM_011516281.2 | 2412 | Silent Mutation | ACA,ACG | T,T 512 | XP_011514583.1 | |
XM_017012263.1 | 2412 | Silent Mutation | ACA,ACG | T,T 485 | XP_016867752.1 |