Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTCACATCTGAAAACATGAGGA[A/G]TGGACTGGTTAATAGTGAAGTCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613413 | ||||||||||||||||||||
Literature Links: |
TMEM106B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM106B - transmembrane protein 106B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134232.1 | 289 | Missense Mutation | AAT,AGT | N,S 28 | NP_001127704.1 | |
NM_018374.3 | 289 | Missense Mutation | AAT,AGT | N,S 28 | NP_060844.2 | |
XM_005249789.1 | 289 | Missense Mutation | AAT,AGT | N,S 28 | XP_005249846.1 |