Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCTGGGGATGACCTTTTCCCC[A/G]AAGATGATAAAGGTAATGCCATCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604707 | ||||||||||||||||||||
Literature Links: |
AKR1B10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR1B10 - aldo-keto reductase family 1 member B10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020299.4 | 693 | Missense Mutation | AAA,GAA | K,E 125 | NP_064695.3 | |
XM_011516416.1 | 693 | Missense Mutation | AAA,GAA | K,E 86 | XP_011514718.1 | |
XM_011516417.2 | 693 | Missense Mutation | AAA,GAA | K,E 125 | XP_011514719.1 |