Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGGATAATTTTTTCAGCCGCT[G/T]TGTGAGCCATTTTCTCTTTTGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM200A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM200A - family with sequence similarity 200 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145111.3 | 420 | Missense Mutation | NP_659802.1 | |||
XM_011515911.2 | 420 | Missense Mutation | XP_011514213.1 | |||
XM_011515913.2 | 420 | Missense Mutation | XP_011514215.1 |
ZNF655 - zinc finger protein 655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |