Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCAGGCTGCCAGGCGGGGCTC[A/G]CCGCCTTCGCAGTGCACGTTGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607639 MIM: 182889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSC4D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSC4D - scavenger receptor cysteine rich family member with 4 domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080744.1 | 80 | Silent Mutation | NP_542782.1 | |||
XM_017011750.1 | 80 | Silent Mutation | XP_016867239.1 |
ZP3 - zona pellucida glycoprotein 3 (sperm receptor) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110354.1 | 80 | Intron | NP_001103824.1 | |||
NM_007155.5 | 80 | UTR 5 | NP_009086.4 | |||
XM_017012586.1 | 80 | UTR 5 | XP_016868075.1 |