Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGCTCCACGGACTCCGGCTC[C/G]CGGTTCTGCGCGGTGGCGCTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600312 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6836 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6836 - microRNA 6836 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT1 - nudix hydrolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013321.3 | 1258 | Silent Mutation | CGC,CGG | R,R 364 | NP_037453.1 | |
XM_011515329.2 | 1258 | Silent Mutation | CGC,CGG | R,R 356 | XP_011513631.1 | |
XM_011515330.1 | 1258 | Silent Mutation | CGC,CGG | R,R 350 | XP_011513632.1 | |
XM_017012083.1 | 1258 | Silent Mutation | CGC,CGG | R,R 311 | XP_016867572.1 | |
XM_017012084.1 | 1258 | Silent Mutation | CGC,CGG | R,R 311 | XP_016867573.1 |