Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAGACCTGGTGTTTCAATACCA[C/G]TGCAGTTACAGACTCTTCCTGGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP4 - Sp4 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326542.1 | 939 | Missense Mutation | CTG,GTG | L,V 224 | NP_001313471.1 | |
NM_001326543.1 | 939 | UTR 5 | NP_001313472.1 | |||
NM_003112.4 | 939 | Missense Mutation | CTG,GTG | L,V 241 | NP_003103.2 | |
XM_005249828.2 | 939 | Missense Mutation | CTG,GTG | L,V 224 | XP_005249885.1 | |
XM_005249829.3 | 939 | Missense Mutation | CTG,GTG | L,V 241 | XP_005249886.1 | |
XM_011515486.2 | 939 | Missense Mutation | CTG,GTG | L,V 241 | XP_011513788.1 | |
XM_011515487.2 | 939 | Missense Mutation | CTG,GTG | L,V 241 | XP_011513789.1 | |
XM_011515489.2 | 939 | Missense Mutation | CTG,GTG | L,V 241 | XP_011513791.1 | |
XM_017012557.1 | 939 | Missense Mutation | CTG,GTG | L,V 241 | XP_016868046.1 |