Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGACCTAAGGGTTGGGCTCATCA[A/G]CTGGGCAGGAACCTACCTCACCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103188 MIM: 615800 MIM: 607418 | ||||||||||||||||||||
Literature Links: |
ARF5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARF5 - ADP ribosylation factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FSCN3 - fascin actin-bundling protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020369.2 | 284 | Missense Mutation | AAC,AGC | N,S 22 | NP_065102.1 |
GCC1 - GRIP and coiled-coil domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |