Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACAATAAAAAGGCAAAAAATT[A/C]TATCAGAAGAACTGCAACAGAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608498 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf72 - chromosome 7 open reading frame 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161834.2 | 700 | Missense Mutation | ATA,CTA | I,L 159 | NP_001155306.2 | |
XM_011515052.1 | 700 | Missense Mutation | ATA,CTA | I,L 159 | XP_011513354.1 | |
XM_011515053.2 | 700 | Missense Mutation | ATA,CTA | I,L 159 | XP_011513355.1 |
ZPBP - zona pellucida binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |