Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGGCCCTCCTGTGAGCGTCGC[A/G]TCAATCACATGGTCGTTATCCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606833 | ||||||||||||||||||||
Literature Links: |
KMT2C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KMT2C - lysine methyltransferase 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170606.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4836 | NP_733751.2 | |
XM_005250025.4 | 14922 | Silent Mutation | GAC,GAT | D,D 4908 | XP_005250082.1 | |
XM_005250026.3 | 14922 | Silent Mutation | GAC,GAT | D,D 4907 | XP_005250083.1 | |
XM_005250027.4 | 14922 | Silent Mutation | GAC,GAT | D,D 4907 | XP_005250084.1 | |
XM_005250028.4 | 14922 | Silent Mutation | GAC,GAT | D,D 4904 | XP_005250085.1 | |
XM_005250031.4 | 14922 | Silent Mutation | GAC,GAT | D,D 4853 | XP_005250088.1 | |
XM_006716077.3 | 14922 | Silent Mutation | GAC,GAT | D,D 4907 | XP_006716140.1 | |
XM_006716078.3 | 14922 | Silent Mutation | GAC,GAT | D,D 4884 | XP_006716141.1 | |
XM_006716079.3 | 14922 | Silent Mutation | GAC,GAT | D,D 4852 | XP_006716142.1 | |
XM_011516450.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4892 | XP_011514752.1 | |
XM_011516451.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4868 | XP_011514753.1 | |
XM_011516452.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4857 | XP_011514754.1 | |
XM_011516453.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4829 | XP_011514755.1 | |
XM_011516454.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4603 | XP_011514756.1 | |
XM_011516456.2 | 14922 | Silent Mutation | GAC,GAT | D,D 4888 | XP_011514758.1 | |
XM_017012480.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4908 | XP_016867969.1 | |
XM_017012481.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4907 | XP_016867970.1 | |
XM_017012482.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4903 | XP_016867971.1 | |
XM_017012483.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4903 | XP_016867972.1 | |
XM_017012484.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4897 | XP_016867973.1 | |
XM_017012485.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4891 | XP_016867974.1 | |
XM_017012486.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4879 | XP_016867975.1 | |
XM_017012487.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4859 | XP_016867976.1 | |
XM_017012488.1 | 14922 | Silent Mutation | GAC,GAT | D,D 4847 | XP_016867977.1 | |
XM_017012489.1 | 14922 | Silent Mutation | GAC,GAT | D,D 3798 | XP_016867978.1 | |
XM_017012490.1 | 14922 | Silent Mutation | GAC,GAT | D,D 3666 | XP_016867979.1 |
LOC731075 - uncharacterized LOC731075 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YBX1P4 - Y-box binding protein 1 pseudogene 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |