Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACATCAACCCACTGCGCCGCTCG[C/T]GCCTAACGCCCAACGACCTGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605464 MIM: 611741 MIM: 123831 MIM: 109280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB8 - ATP binding cassette subfamily B member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ASIC3 - acid sensing ion channel subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004769.3 | 895 | Missense Mutation | CGC,TGC | R,C 101 | NP_004760.1 | |
NM_020321.3 | 895 | Missense Mutation | CGC,TGC | R,C 101 | NP_064717.1 | |
NM_020322.3 | 895 | Missense Mutation | CGC,TGC | R,C 101 | NP_064718.1 |
CDK5 - cyclin dependent kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A2 - solute carrier family 4 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |