Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCGCTGAAGAGGCTGTCACTCT[C/T]CTCGAAGATCTGGAGCGGGAACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194510 MIM: 601261 | ||||||||||||||||||||
Literature Links: |
ZNF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF3 - zinc finger protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN21 - zinc finger and SCAN domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145914.2 | 501 | Silent Mutation | CTC,CTT | L,L 119 | NP_666019.1 | |
XM_005250568.4 | 501 | Silent Mutation | CTC,CTT | L,L 119 | XP_005250625.1 | |
XM_006716113.3 | 501 | Silent Mutation | CTC,CTT | L,L 119 | XP_006716176.1 | |
XM_017012584.1 | 501 | Silent Mutation | CTC,CTT | L,L 119 | XP_016868073.1 | |
XM_017012585.1 | 501 | Silent Mutation | CTC,CTT | L,L 119 | XP_016868074.1 |