Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACCAACCACAAAAAAGTGGCAA[C/G]AAGTGTAACAACAAAGTCCTAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYPL1 - synaptophysin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006754.3 | 593 | Missense Mutation | CTT,GTT | L,V 159 | NP_006745.1 | |
NM_182715.2 | 593 | Missense Mutation | CTT,GTT | L,V 141 | NP_874384.1 |