Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTATCTTTCTTCTTGCATATGG[C/T]TGACATTTTAATTAAGATGTTAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISPD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISPD - isoprenoid synthase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101417.3 | 5228 | UTR 3 | NP_001094887.1 | |||
NM_001101426.3 | 5228 | UTR 3 | NP_001094896.1 | |||
XM_006715770.3 | 5228 | Intron | XP_006715833.1 | |||
XM_011515499.2 | 5228 | Intron | XP_011513801.1 | |||
XM_011515500.2 | 5228 | Intron | XP_011513802.1 | |||
XM_011515501.2 | 5228 | Intron | XP_011513803.1 | |||
XM_011515508.2 | 5228 | Intron | XP_011513810.1 | |||
XM_011515509.2 | 5228 | Intron | XP_011513811.1 | |||
XM_017012575.1 | 5228 | Intron | XP_016868064.1 | |||
XM_017012576.1 | 5228 | Intron | XP_016868065.1 | |||
XM_017012577.1 | 5228 | Intron | XP_016868066.1 | |||
XM_017012578.1 | 5228 | Intron | XP_016868067.1 |