Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTTTAGAATCACGAAAGCTTA[A/G]CTGCTGTGGTTTAAAGTACAGTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608491 MIM: 616993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMTF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMTF1 - cyclin D binding myb like transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142326.1 | 4111 | UTR 3 | NP_001135798.1 | |||
NM_001142327.1 | 4111 | UTR 3 | NP_001135799.1 | |||
NM_021145.3 | 4111 | UTR 3 | NP_066968.3 | |||
XM_011516735.2 | 4111 | UTR 3 | XP_011515037.1 | |||
XM_011516737.1 | 4111 | UTR 3 | XP_011515039.1 | |||
XM_011516739.2 | 4111 | UTR 3 | XP_011515041.1 | |||
XM_017012865.1 | 4111 | UTR 3 | XP_016868354.1 | |||
XM_017012866.1 | 4111 | UTR 3 | XP_016868355.1 | |||
XM_017012867.1 | 4111 | UTR 3 | XP_016868356.1 | |||
XM_017012868.1 | 4111 | UTR 3 | XP_016868357.1 | |||
XM_017012869.1 | 4111 | UTR 3 | XP_016868358.1 | |||
XM_017012870.1 | 4111 | UTR 3 | XP_016868359.1 | |||
XM_017012871.1 | 4111 | UTR 3 | XP_016868360.1 | |||
XM_017012872.1 | 4111 | UTR 3 | XP_016868361.1 | |||
XM_017012873.1 | 4111 | UTR 3 | XP_016868362.1 | |||
XM_017012874.1 | 4111 | UTR 3 | XP_016868363.1 | |||
XM_017012875.1 | 4111 | UTR 3 | XP_016868364.1 | |||
XM_017012876.1 | 4111 | UTR 3 | XP_016868365.1 | |||
XM_017012877.1 | 4111 | UTR 3 | XP_016868366.1 | |||
XM_017012878.1 | 4111 | UTR 3 | XP_016868367.1 |
TMEM243 - transmembrane protein 243 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005250586.3 | 4111 | Intron | XP_005250643.1 | |||
XM_005250587.4 | 4111 | Intron | XP_005250644.1 | |||
XM_005250588.3 | 4111 | Intron | XP_005250645.1 | |||
XM_005250589.3 | 4111 | Intron | XP_005250646.1 | |||
XM_017012616.1 | 4111 | Intron | XP_016868105.1 | |||
XM_017012617.1 | 4111 | Intron | XP_016868106.1 | |||
XM_017012618.1 | 4111 | Intron | XP_016868107.1 |