Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGTCCTGTGGGCGGTCAGCGT[A/G]GATCCAGTGGCCAGCGTTCGGCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612545 | ||||||||||||||||||||
Literature Links: |
ABHD11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD11 - abhydrolase domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145364.2 | 1062 | Missense Mutation | CAC,TAC | H,Y 242 | NP_001138836.1 | |
NM_001301058.1 | 1062 | UTR 3 | NP_001287987.1 | |||
NM_001321382.1 | 1062 | Missense Mutation | CAC,TAC | H,Y 186 | NP_001308311.1 | |
NM_001321383.1 | 1062 | UTR 3 | NP_001308312.1 | |||
NM_148912.3 | 1062 | Missense Mutation | CAC,TAC | H,Y 299 | NP_683710.1 | |
NM_148913.3 | 1062 | Missense Mutation | CAC,TAC | H,Y 292 | NP_683711.1 | |
XM_006716144.2 | 1062 | Missense Mutation | CAC,TAC | H,Y 186 | XP_006716207.1 | |
XM_011516590.2 | 1062 | Missense Mutation | CAC,TAC | H,Y 278 | XP_011514892.1 | |
XM_011516591.1 | 1062 | Missense Mutation | CAC,TAC | H,Y 186 | XP_011514893.1 |
ABHD11-AS1 - ABHD11 antisense RNA 1 (tail to tail) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |