Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTCGAGTAGTGGCGCTAGAGGC[A/G]GCCTGGCCCTGTGGGCAGGACCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616813 MIM: 606965 MIM: 109280 MIM: 614792 | ||||||||||||||||||||
Literature Links: |
AGAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGAP3 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FASTK - Fas activated serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258461.1 | 1064 | Silent Mutation | GCC,GCT | A,A 437 | NP_001245390.1 | |
NM_006712.4 | 1064 | Silent Mutation | GCC,GCT | A,A 464 | NP_006703.1 | |
NM_033015.3 | 1064 | Silent Mutation | GCC,GCT | A,A 323 | NP_148936.2 | |
XM_005249932.1 | 1064 | Silent Mutation | GCC,GCT | A,A 430 | XP_005249989.1 | |
XM_005249933.3 | 1064 | Silent Mutation | GCC,GCT | A,A 430 | XP_005249990.1 | |
XM_011515761.2 | 1064 | Silent Mutation | GCC,GCT | A,A 464 | XP_011514063.1 | |
XM_011515762.2 | 1064 | Intron | XP_011514064.1 | |||
XM_011515763.1 | 1064 | Silent Mutation | GCC,GCT | A,A 296 | XP_011514065.1 | |
XM_017011704.1 | 1064 | Silent Mutation | GCC,GCT | A,A 403 | XP_016867193.1 | |
XM_017011705.1 | 1064 | Silent Mutation | GCC,GCT | A,A 323 | XP_016867194.1 | |
XM_017011706.1 | 1064 | Silent Mutation | GCC,GCT | A,A 225 | XP_016867195.1 |
SLC4A2 - solute carrier family 4 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199692.1 | 1064 | Intron | NP_001186621.1 | |||
NM_001199693.1 | 1064 | Intron | NP_001186622.1 | |||
NM_001199694.1 | 1064 | Intron | NP_001186623.1 | |||
NM_003040.3 | 1064 | Intron | NP_003031.3 | |||
XM_006716094.3 | 1064 | Intron | XP_006716157.1 |
TMUB1 - transmembrane and ubiquitin like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |