Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAATTAAATGGAAAATTTCCGT[C/G]CCTTTTAAGTTACCAATCATAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609978 | ||||||||||||||||||||
Literature Links: |
CADPS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF133 - ring finger protein 133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139175.1 | 716 | Silent Mutation | GGC,GGG | G,G 160 | NP_631914.1 |
RNF148 - ring finger protein 148 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |