Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGTGTGGGCCACATCAGCGGC[A/G]CCCACCTCAACCCGGCTGTCACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107776 | ||||||||||||||||||||
Literature Links: |
AQP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP1 - aquaporin 1 (Colton blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185060.1 | Intron | NP_001171989.1 | ||||
NM_001185061.1 | Intron | NP_001171990.1 | ||||
NM_001185062.1 | Intron | NP_001171991.1 |