Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGATCCTCCATGCTGCTGTAT[C/T]GGAAAATGTCACTCACGTTCTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602981 MIM: 600815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AEBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AEBP1 - AE binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4649 - microRNA 4649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLD2 - DAN polymerase delta 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127218.2 | 1711 | Missense Mutation | CAA,CGA | Q,R 354 | NP_001120690.1 | |
NM_001256879.1 | 1711 | Missense Mutation | CAA,CGA | Q,R 354 | NP_001243808.1 | |
NM_006230.3 | 1711 | Missense Mutation | CAA,CGA | Q,R 389 | NP_006221.2 | |
XM_011515431.1 | 1711 | Missense Mutation | CAA,CGA | Q,R 387 | XP_011513733.1 |