Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAAATTTATTTTTTCAATGAA[C/T]GTTCTTAGGCATGTGTAAGAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK6 - cyclin dependent kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145306.1 | 5870 | UTR 3 | NP_001138778.1 | |||
NM_001259.6 | 5870 | UTR 3 | NP_001250.1 | |||
XM_006715835.2 | 5870 | UTR 3 | XP_006715898.1 |