Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAGCACCTCGTGAACCACTACT[A/G]CTGCAAGCGCCCCATCTATGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611725 | ||||||||||||||||||||
Literature Links: |
KCTD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD7 - potassium channel tetramerization domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167961.2 | 1011 | Missense Mutation | TAC,TGC | Y,C 276 | NP_001161433.1 | |
NM_153033.4 | 1011 | Missense Mutation | TAC,TGC | Y,C 276 | NP_694578.1 |