Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCAGGGACTTCTGGAAAAGATC[A/G]CAACGCCAAAGGTGAAGAACCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602321 | ||||||||||||||||||||
Literature Links: |
GSTK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTK1 - glutathione S-transferase kappa 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143679.1 | 434 | Missense Mutation | ACA,GCA | T,A 216 | NP_001137151.1 | |
NM_001143680.1 | 434 | Missense Mutation | ACA,GCA | T,A 148 | NP_001137152.1 | |
NM_001143681.1 | 434 | Missense Mutation | ACA,GCA | T,A 117 | NP_001137153.1 | |
NM_015917.2 | 434 | Missense Mutation | ACA,GCA | T,A 160 | NP_057001.1 |
LOC105375545 - uncharacterized LOC105375545 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |