Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTAGAGGGGGTTAATGTGCAGC[A/G]TTTCAGACTAGAAGAGAGAATTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf73 - chromosome 7 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318192.1 | 2324 | Missense Mutation | ACG,ATG | T,M 539 | NP_001305121.1 | |
NM_012450.3 | 2324 | Missense Mutation | ACG,ATG | T,M 538 | NP_036582.2 | |
XM_011516024.2 | 2324 | Intron | XP_011514326.1 | |||
XM_017011962.1 | 2324 | Missense Mutation | ACG,ATG | T,M 487 | XP_016867451.1 | |
XM_017011963.1 | 2324 | Intron | XP_016867452.1 |