Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGCCCGAGAAGCTGATGGGCTG[A/G]CAGGCCAGGCTGCGCAGGTTCACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602981 MIM: 600815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AEBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AEBP1 - AE binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129.4 | 1991 | Intron | NP_001120.3 | |||
XM_011515162.1 | 1991 | Intron | XP_011513464.1 |
MIR4649 - microRNA 4649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLD2 - DAN polymerase delta 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127218.2 | 1991 | Silent Mutation | NP_001120690.1 | |||
NM_001256879.1 | 1991 | Silent Mutation | NP_001243808.1 | |||
NM_006230.3 | 1991 | Silent Mutation | NP_006221.2 | |||
XM_011515431.1 | 1991 | Silent Mutation | XP_011513733.1 |