Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGAGCACAGCCCCGAAGCCAT[C/T]TTTGTCCTGAAGTGTCTTTAGTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO1G - myosin IG | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033054.2 | 2493 | Missense Mutation | AAT,GAT | N,D 864 | NP_149043.2 | |
XM_017012503.1 | 2493 | Missense Mutation | AAT,GAT | N,D 749 | XP_016867992.1 | |
XM_017012504.1 | 2493 | Missense Mutation | AAT,GAT | N,D 626 | XP_016867993.1 |