Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGGGGATCTTGAGGTTTTCAG[G/T]TGAACAGATTCAAAATGGTCTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614048 | ||||||||||||||||||||
Literature Links: |
KIAA1324L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1324L - KIAA1324 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142749.2 | 3245 | Silent Mutation | NP_001136221.1 | |||
NM_001291990.1 | 3245 | Silent Mutation | NP_001278919.1 | |||
NM_001291991.1 | 3245 | Silent Mutation | NP_001278920.1 | |||
NM_152748.3 | 3245 | Silent Mutation | NP_689961.3 | |||
XM_006715894.3 | 3245 | Silent Mutation | XP_006715957.1 | |||
XM_011515918.2 | 3245 | Silent Mutation | XP_011514220.1 | |||
XM_011515921.1 | 3245 | Missense Mutation | XP_011514223.1 | |||
XM_017011843.1 | 3245 | Missense Mutation | XP_016867332.1 | |||
XM_017011844.1 | 3245 | Silent Mutation | XP_016867333.1 |