Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCCTTCGCTTCCCTGCATCTGC[A/G]CTGATTGGTAAGTGCTTCAGATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602987 MIM: 604088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE1C - phosphodiesterase 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191056.3 | 115 | Intron | NP_001177985.1 | |||
NM_001191057.3 | 115 | Intron | NP_001177986.1 | |||
NM_001191058.3 | 115 | Intron | NP_001177987.2 | |||
NM_001191059.3 | 115 | Intron | NP_001177988.1 | |||
NM_001322055.1 | 115 | Intron | NP_001308984.1 | |||
NM_001322056.1 | 115 | Intron | NP_001308985.1 | |||
NM_001322057.1 | 115 | Intron | NP_001308986.1 | |||
NM_001322058.1 | 115 | Intron | NP_001308987.1 | |||
NM_001322059.1 | 115 | Intron | NP_001308988.1 | |||
NM_005020.4 | 115 | Intron | NP_005011.1 | |||
XM_017012264.1 | 115 | Intron | XP_016867753.1 | |||
XM_017012265.1 | 115 | Intron | XP_016867754.1 | |||
XM_017012266.1 | 115 | Intron | XP_016867755.1 | |||
XM_017012267.1 | 115 | Intron | XP_016867756.1 |
PPP1R17 - protein phosphatase 1 regulatory subunit 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145123.2 | 115 | UTR 5 | NP_001138595.1 | |||
NM_006658.4 | 115 | UTR 5 | NP_006649.2 | |||
XM_011515094.1 | 115 | UTR 5 | XP_011513396.1 |