Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTATCAATAGAAATAAGGATAT[C/T]ATGAAGATTGGTTGCTCACTGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605790 | ||||||||||||||||||||
Literature Links: |
STK31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STK31 - serine/threonine kinase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260504.1 | 219 | Silent Mutation | ATC,ATT | I,I 35 | NP_001247433.1 | |
NM_001260505.1 | 219 | Silent Mutation | ATC,ATT | I,I 58 | NP_001247434.1 | |
NM_031414.4 | 219 | Silent Mutation | ATC,ATT | I,I 58 | NP_113602.2 | |
NM_032944.3 | 219 | Silent Mutation | ATC,ATT | I,I 35 | NP_116562.2 | |
XM_006715758.3 | 219 | Silent Mutation | ATC,ATT | I,I 58 | XP_006715821.1 | |
XM_011515449.2 | 219 | Silent Mutation | ATC,ATT | I,I 62 | XP_011513751.2 | |
XM_011515450.1 | 219 | Silent Mutation | ATC,ATT | I,I 35 | XP_011513752.1 | |
XM_011515452.2 | 219 | Silent Mutation | ATC,ATT | I,I 62 | XP_011513754.2 |