Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCTGTGGACAGTGGTGGCCGC[C/T]GTGCAGGCTATAGAGAGGAAGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF398 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF398 - zinc finger protein 398 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020781.3 | 258 | UTR 5 | NP_065832.1 | |||
NM_170686.2 | 258 | Silent Mutation | GCC,GCT | A,A 49 | NP_733787.1 | |
XM_011516439.2 | 258 | Silent Mutation | GCC,GCT | A,A 54 | XP_011514741.1 | |
XM_011516440.2 | 258 | Silent Mutation | GCC,GCT | A,A 42 | XP_011514742.1 | |
XM_011516441.2 | 258 | UTR 5 | XP_011514743.1 |