Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTTGGATTGCAAAGTCTATACC[A/G]GACTGCAGCCGTGTAATAAGCATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM200A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM200A - family with sequence similarity 200 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145111.3 | 592 | Silent Mutation | TCC,TCT | S,S 136 | NP_659802.1 | |
XM_011515911.2 | 592 | Silent Mutation | TCC,TCT | S,S 136 | XP_011514213.1 | |
XM_011515913.2 | 592 | Silent Mutation | TCC,TCT | S,S 136 | XP_011514215.1 |
ZNF655 - zinc finger protein 655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |