Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACTGTCTTGGCTTTTGTCCAT[A/G]CTACTATTGTCGCTTGCCTGGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGK - acylglycerol kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018238.3 | Intron | NP_060708.1 | ||||
XM_005250023.3 | Intron | XP_005250080.1 | ||||
XM_011516397.2 | Intron | XP_011514699.1 |