Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACACGATTGGAGTGGACTTTAC[C/T]GTGCGTTCCCTTGATATTGACGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RAB19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB19 - RAB19, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008749.2 | 559 | Silent Mutation | ACC,ACT | T,T 55 | NP_001008749.2 | |
XM_005249996.3 | 559 | Silent Mutation | ACC,ACT | T,T 55 | XP_005250053.1 | |
XM_005249997.3 | 559 | Silent Mutation | ACC,ACT | T,T 55 | XP_005250054.1 | |
XM_005249998.3 | 559 | Silent Mutation | ACC,ACT | T,T 55 | XP_005250055.1 | |
XM_017012207.1 | 559 | Silent Mutation | ACC,ACT | T,T 55 | XP_016867696.1 |
SLC37A3 - solute carrier family 37 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |