Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGATGAGGGAGAGAGGCACAA[C/T]GAAGCAGAAGATGAAGAGGAACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603420 MIM: 613522 | ||||||||||||||||||||
Literature Links: |
CALU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALU - calumenin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPN1SW - opsin 1 (cone pigments), short-wave-sensitive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001708.2 | 628 | Missense Mutation | ATT,GTT | I,V 210 | NP_001699.1 |