Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGCGCAGCCTGTAACTTCAGCA[A/G]CATTCTAGTGAATCAGGAGCGGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EEPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEPD1 - endonuclease/exonuclease/phosphatase family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030636.2 | 813 | Missense Mutation | AAC,AGC | N,S 32 | NP_085139.2 | |
XM_017012657.1 | 813 | Missense Mutation | AAC,AGC | N,S 32 | XP_016868146.1 | |
XM_017012658.1 | 813 | Missense Mutation | AAC,AGC | N,S 32 | XP_016868147.1 |