Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGGCCTTGTGAATGCTCAGTCG[A/G]AGGTGATGAATTATCTACAAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZC3HAV1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZC3HAV1L - zinc finger CCCH-type containing, antiviral 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080660.3 | 1132 | Missense Mutation | CCG,TCG | P,S 259 | NP_542391.2 | |
XM_006716176.3 | 1132 | UTR 3 | XP_006716239.1 | |||
XM_011516688.2 | 1132 | Missense Mutation | CCG,TCG | P,S 259 | XP_011514990.1 | |
XM_011516689.2 | 1132 | Missense Mutation | CCG,TCG | P,S 123 | XP_011514991.1 |